Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SIRPA Rabbit Polyclonal Antibody

Supplier: Genetex

GTX28120
89362-728EA 430.75 USD InStock
89362-728
Anti-SIRPA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to SIRP alpha

Western blot at1:1000 to 1:2000 dilution, and 75-110 kDa bandsshould be detected.

Type: Primary
Antigen: SIRPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty