Anti-ACT1 Mouse Monoclonal Antibody

Supplier: GeneTex
GTX27799
89352-302EA 520.82 USD
89352-302
Anti-ACT1 Mouse Monoclonal Antibody
Antibodies
This antibody labels skeletal and cardiac muscle tissue.

Type: Primary
Antigen: ACT1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Carp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR