Anti-S100 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX73545
89353-204EA 585.96 USD
89353-204
Anti-S100 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with cow S-100 protein A and B. It shows strong cross-reactivity with human S-100 A and B. In the brain, it labels glial and ependymal cells. Melanocytes and Langerhans cells of the skin stain positive with this antibody.

Type: Primary
Antigen: S100
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR