Anti-ENO2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX15488
89351-130EA 387.41 USD
89351-130
Anti-ENO2 Rabbit Polyclonal Antibody
Antibodies
Enolase is a glycolytic enzyme catalyzing the reaction pathway between 2-phospho-glycerate and phosphoenol pyruvate. In mammals, enolase molecules are dimers composed of three distinct subunits (alpha, beta and gamma). The alpha subunit is expressed in most tissues and the beta subunit only in muscle. The gamma subunit is expressed primarily in neurons, in normal and in neoplastic neuroendocrine cells. Coexpression of NSE (neuron specific enolase) and chromogranin A is common in neuroendocrine neoplasms.

IHC-P: Working dilution, ready-to-use for 10 minutes at RT. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Citrate buffer, pH 6.0 for 10 minutes, followed by cooling at RT for 20 minutes. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ENO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR