Anti-ENO2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX72911
89353-076EA 624.33 USD
89353-076
Anti-ENO2 Rabbit Polyclonal Antibody
Antibodies
Enolase is a glycolytic enzyme catalyzing the reaction pathway between 2- phospho-glycerate and phosphoenol pyruvate. In mammals, enolase molecules are dimers composed of three distinct subunits (alpha, beta and gamma). The alpha subunit is expressed in most tissues and the beta subunit only in muscle. The gamma subunit is expressed primarily in neurons, in normal and in neoplastic neuroendocrine cells. Coexpression of NSE (neuron specific enolase) and chromogranin A is common in neuroendocrine neoplasms.

Type: Primary
Antigen: ENO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR