Anti-JNK2 Rabbit Polyclonal Antibody

GTX19330
89367-506EA 456.31 USD
89367-506
Anti-JNK2 Rabbit Polyclonal Antibody
c-Jun amino terminal kinases (JNKs) are MAP kinase family members that differ from other MAP kinases in that they are phosphorylated at the Thr-Pro-Tyr phosphorylation motif instead of the characteristic MAP kinase Thr-Glu-Tyr motif. JNK2 (p54a, SAPK1a), along with JNK1 and JNK3, is thought to play an important role in nuclear signal transduction through its environmental stress activation and subsequent phosphorylation of the nuclear transcription factor p53.

For WB: Use at a dilution of 1:500. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: JNK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR