Anti-JNK1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX27949
89352-320EA 602.26 USD
89352-320
Anti-JNK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to JAK1

For IHC: Use at an assay dependent dilution. For IP: Use at an assay dependent dilution. For WB: Use at a dilution of 1:200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: JNK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR