Goat polyclonal to IL2
ELISA: Use at a concentration of 1 ?g/ml, this detects 0.6 ng/well of recombinant, mouse IL2. Neut: Use at a concentration of 3.5 ?g/ml. WB: Use at a concentration of 2 ?g/ml, this detects 5 ng/lane of recombinant, mouse IL2 under reducing and non-reducing conditions. Predicted molecular weight: 19 kDa. Anti-Mouse IL2 is tested for its ability to neutralize the biological activity of recombinant, mouse IL2 on CTLL2 cells. The ND50 of the antibody is defined as the concentration of antibody resulting in a one-half maximal inhibition of bioactivity of recombinant, mouse IL2, when IL2 is present at a concentration just high enough to elicit a maximum response. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: IL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse