Anti-NFKBIA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX27545
89362-604EA 606.86 USD
89362-604
Anti-NFKBIA Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to IKB alpha

For ELISA: Use at an assay dependant dilution. For IP: Use at an assay dependant dilution. For WB: Use at a dilution of 1:1,000. Detects a band of approximately 36 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: NFKBIA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR