Anti-IGF2 Goat Polyclonal Antibody

Supplier: GeneTex
GTX10731
89354-908EA 567.96 USD
89354-908
Anti-IGF2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to IGF2

ELISA: Use at a concentration of 0.5 - 1.0 ?g/ml. Neut: Use at a concentration of 1.5 - 4.5 ?g/ml. WB: Use at a concentration of 0.1 - 0.2 ?g/ml. Predicted molecular weight: 20 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IGF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR