Anti-IgG Sheep Polyclonal Antibody

Supplier: GeneTex
GTX26866
89362-210EA 240.87 USD
89362-210
Anti-IgG Sheep Polyclonal Antibody
Antibodies
FITC-conjugated Sheep anti-Human IgG polyclonal antibody

Recommended Starting Dilutions: Not specifiedOptimal titers for applications should be determined by the researcher.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR