Anti-GST Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX19564
89351-486EA 784.57 USD
89351-486
Anti-GST Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR