Anti-EB3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX13859
89359-872EA 602.26 USD
89359-872
Anti-EB3 Rabbit Polyclonal Antibody
Antibodies
EB3 is a 282 amino acid homologue of EB1, a protein known to bind Adenomatous Polyposis Coli. It has been shown to bind to tubulin in vitro and microtubules in vivo. Northern blotting shows expressions to be mainly in brain, and to a lesser degree in muscle. EB3 interacts with APCL, a homologue of the tumor suppressor APC, and this interaction may be involved in the stabilization or extension of microtubules.

WB: Use at a concentration of 0.5 - 2 ?g/ml. Detects a band of approximately 38 kDa (predicted molecular weight: 34 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: EB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR