Anti-CD68 Mouse Monoclonal Antibody [clone: 514H12]

Supplier: Genetex

GTX75390
89365-784EA 209.27 USD
89365-784
Anti-CD68 Mouse Monoclonal Antibody [clone: 514H12]
Antibodies
Mouse monoclonal antibody [514H12] to CD68

Recommended Starting Dilutions: Immunohistology - Paraffin: 1/40 - 1/80 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: 514H12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR