Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

Anti-CD4 Mouse Monoclonal Antibody

Supplier: Genetex

GTX76321
89366-054EA 460.43 USD InStock
89366-054
Anti-CD4 Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal [W3/25] to rat CD4

  • For FACS: Use at a dilution of 1:100 - 1:200. For IHC-Fr and IHC-P: Use at an assay dependent dilution. For Functional studies: Use at an assay dependent solution. Not tested in other applications. Optimal concentrations/dilutions should be determined by the reseacher.Immunohistology: *This clone has been described reacting with paraffin-embedded material following PLP fixation (periodate-lysine-paraformaldehyde)

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty