Anti-CD1A Mouse Monoclonal Antibody [clone: NA1/34-HLK]

Supplier: GeneTex
GTX76538
89367-994EA 479.61 USD
89367-994
Anti-CD1A Mouse Monoclonal Antibody [clone: NA1/34-HLK]
Mouse Antibody to Human CD1a:Azide Free

For Flow Cytometry: Use at a dilution of 1:50 - 1:100. Use 10ul of the suggested working dilution to label 10^6 cells in 100ul. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR