Anti-CCL4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX45129
89363-574EA 660.25 USD
89363-574
Anti-CCL4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to CCL4

For WB: Use at a dilution of 1:2,000. For IP: Use at a dilution of 1:500. The use of this product can also be used, but not yet reported. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CCL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR