Anti-BRCA1 Mouse Polyclonal Antibody

Supplier: GeneTex
GTX70134
89364-700EA 583.47 USD
89364-700
Anti-BRCA1 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antiserum to BRCA1

For IP: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Predicted molecular weight: 208 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: BRCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR