Anti-BRCA1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX22956
89351-834EA 377 USD
89351-834
Anti-BRCA1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to BRCA1

Immunofluorescence: assay dependent. Immunoprecipitation (with protein A): use at a concentration of 10?g / mg of lysate (co-immunoprecipitates EGFR). Western Blotting: use at a concentration of 15 - 30?g / ml for 2h at RT. Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. By Western blot, this antibody detects a band of 220 kDa, which corresponds to the predicted molecular weight of BRCA 1.

Type: Primary
Antigen: BRCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR