Monoclonal Mouse Antibody to bcl - X ( Apoptosis Marker )
This antibody may be diluted to a titer of 1:25-1:50 in an ABC method. However, depending on the staining conditions employed, we suggest that final dilution should be determined by the user. We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation should be determined by the user. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining. Can also be used for: Immunoprecipitation (1:50), Western Blotting (1:50 to 1:100)
Type: Primary
Antigen: BCL2L
Clonality: Monoclonal
Clone: 2H12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human