Anti-V5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX77434
89353-704EA 628.47 USD
89353-704
Anti-V5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: V5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR