Anti-TRKA Rabbit Polyclonal Antibody

GTX21445
89361-038EA 386.57 USD
89361-038
Anti-TRKA Rabbit Polyclonal Antibody
Trk A, the high affinity Nerve Growth Factor (NGF) receptor, autophosphorylates on tyrosine to activate multiple effectors. Phosphorylation at Tyr490 is required for Shc association and activation of the Ras-MAP kinase cascade. Residues Tyr674/675 lie within the catalytic domain, and phosphorylation at these sites reflects Trk kinase activity.

For ICC: Use at a concentration of 10-20 ?g/ml. For IP: Use at a concentration of 4-8 ?g/ml. For WB: Use at a concentration of 1 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: TRKA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR