Anti-AFM Goat Polyclonal Antibody

Supplier: GeneTex
GTX88333
89366-960EA 578.23 USD
89366-960
Anti-AFM Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Afamin / alpha Albumin

Approx 80kDa band observed in Human Brain (Cerebellum and Cerebral Cortex) lysates (calculated MW of 69.1kDa according to NP_001124.1). Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: AFM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR