Anti-MLANA Mouse Monoclonal Antibody [clone: A103]

GTX75784
89365-902EA 471.29 USD
89365-902
Anti-MLANA Mouse Monoclonal Antibody [clone: A103]
Mouse monoclonal antibody [A103] to Melan A

Type: Primary
Antigen: MLANA
Clonality: Monoclonal
Clone: A103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR