Anti-PTH Mouse Monoclonal Antibody [clone: IE6]

Supplier: GeneTex
GTX48875
89352-898EA 592.61 USD
89352-898
Anti-PTH Mouse Monoclonal Antibody [clone: IE6]
Antibodies
Type: Primary
Antigen: PTH
Clonality: Monoclonal
Clone: IE6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR