Anti-PDE2A Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX14605
89367-338EA 519.38 USD
89367-338
Anti-PDE2A Rabbit Polyclonal Antibody
Antibodies
Cyclic nucleotides are important intracellular second messengers which play an important role in a variety of signal transduction process. The cyclic nucleotides are hydrolyzed and compartmentalized by a family of enzymes called phosphodiesterases. One of the many phosphodiesterases that compartmentalize and hydrolyze cAMP and cGMP into AMP and GMP is PDE2. Two genes, PDE2A and PDE2B, have been identified and cloned and they both have an apparent molecular weight of 100 kDA. PDE2A has specificity for cAMP and its activity is modulated by the presence of cGMP. In contrast, PDE2B does not hydrolyze cGMP nor its activity is altered by the presence of cGMP. Western blot analyses of PDE2A reveals that it is present in a variety of tissues including neocortex, cerebellum, heart, kidney lungs, pulmonary artery and skeletal muscle.

IP: Use at a dilution of 1/250. WB: Use at a dilution of 1/500. Detects a band of approximately 95-97 kDa in samples from rat brain. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PDE2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR