Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PMAIP1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX85521
89366-880EA 370.07 USD InStock
89366-880
Anti-PMAIP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Noxa

Recommended Starting Dilutions: For WB: Use at a concentration of 0.5-2 μg/mL. For IHC: Use at a concentration of 1 μg/mL. For ICC/IF: Use at a concentration of 10 μg/mL. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PMAIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty