Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HIST1H2BD Rabbit Polyclonal Antibody

Supplier: Genetex

GTX21790
89367-538EA 476.58 USD InStock
89367-538
Anti-HIST1H2BD Rabbit Polyclonal Antibody
Antibodies
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fibre is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures.

ICC/IF: Use at a concentration of 0.5 μg/ml. ChIP: Use at a concentration of 2-3 μg/ml. Every new batch of this antibody is tested at Abcam in ChIP.WB: Use at a concentration of 0.1 μg/ml. Detects a band of approximately 17 kDa (predicted molecular weight: 14 kDa).

Type: Primary
Antigen: HIST1H2BD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human, Rat, Arabidopsis, Mammals

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty