Anti-GPCR GPR35 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX78059
89366-490EA 481.52 USD
89366-490
Anti-GPCR GPR35 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to G Protein-Coupled Receptor GPR35

Type: Primary
Antigen: GPR35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR