Anti-P57KIP2 Rabbit Polyclonal Antibody

GTX73128
89353-148EA 376.85 USD
89353-148
Anti-P57KIP2 Rabbit Polyclonal Antibody
Rabbit Polyclonal to p57Kip2

For IHC-P: Use at a dilution of 1:1000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: P57KIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR