Anti-NR4A3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX78011
89366-476EA 590.3 USD
89366-476
Anti-NR4A3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Nuclear Receptor NOR-1 (NR4A3)

Type: Primary
Antigen: NR4A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR