Anti-F2RL3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX70759
89365-100EA 481.52 USD
89365-100
Anti-F2RL3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal to Human Proteinase - Activated Receptor 4

For IHC-P: Use at an asssay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: F2RL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR