Anti-EGF Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX59830
89363-742EA 528.9 USD
89363-742
Anti-EGF Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to EGF

Western blot analysis (0.5-4 µg/ml). Recombinant human EGF (Cat.# 4022-10) can be used as a positive control. Per customer’s feedback, it can also be used in Neutralization and Elisa. However, the optimal conditions should be determined individually.

Type: Primary
Antigen: EGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR