Anti-EGF Goat Polyclonal Antibody

Supplier: GeneTex
GTX74093
89365-382EA 674.69 USD
89365-382
Anti-EGF Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to EGF

Recommended Starting Dilutions: ELISA: Max Dilution 0.5 ug/ml Western Blotting: 0.1 ug/ml - 0.2 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: EGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR