Anti-C-MYC Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX19312
89360-598EA 576.8 USD
89360-598
Anti-C-MYC Rabbit Polyclonal Antibody
Antibodies
The c-Myc protein is a transcription factor, which is encoded by the c-Myc gene on human chromosome 8q24. c-Myc is commonly activated in a variety of tumor cells and plays an important role in cellular proliferation, differentiation, apoptosis and cell cycle progression. The phosphorylation of c-Myc has been investigated and previous studies have suggested a functional association between phosphorylation at Thr58/Ser62 by glycogen synthase kinase 3, cyclin-dependent kinase, ERK2 and C-Jun N-terminal Kinase (JNK) in cell proliferation and cell cycle regulation. Studies also have shown that c-Myc is essential for tumor cell development in vasculogenesis and angiogenesis that distribute blood throughout the cells, and which brought extensive attention in the development of new therapeutic approach for cancer treatment.

ELISA: 1:1,0000 - 1:100,000. ICC: 1:200 - 1:500. WB: Colorimetric: 1:1,000 - 1:10,000. Chemiluminescent: 1:1,000 - 1:30,000. Predicted molecular weight: 49 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: C-MYC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR