Anti-LEP Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX22095
89361-152EA 660.25 USD
89361-152
Anti-LEP Rabbit Polyclonal Antibody
Antibodies
Obesity associated leptin is the product of the OB gene and has been identified with Type II diabetes.

ELISA (suggested working dilution 1:2,400) and immunoblotting (suggested working dilution 1:200 to 1:400). Reactivity in other immunoassays is unknown. Optimal titers for other applications should be determined by the researcher.

Type: Primary
Antigen: LEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR