Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S100A9 Mouse Monoclonal Antibody [clone: MAC387]

Supplier: Genetex

GTX76577
89368-004EA 449.43 USD InStock
89368-004
Anti-S100A9 Mouse Monoclonal Antibody [clone: MAC387]
Antibodies
Mouse monoclonal antibody [MAC387] to Macrophage + Monocyte

Recommended Starting Dilutions: Immunohistology - Frozen: 1/100 - 1/200 Flow Cytometry: 1/50 - 1/100 Immunohistology - Paraffin: 1/100 - 1/200 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: S100A9
Clonality: Monoclonal
Clone: MAC387
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog, Pig, Rat, Bb, Cat, Deer, Goat, Guinea Pig, Horse, Rabbit, Bovine, Rhesus Monkey, ClMonkey

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty