Anti-IL6 Goat Polyclonal Antibody

Supplier: Genetex

GTX29770
89362-844EA 400.23 USD
89362-844
Anti-IL6 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to IL6

For ELISA: By direct ELISA (using 100 µl/well antibody solution) Use at a concentration of 0.5 µg/ml. In conjunction with compatible secondary reagents, allows the detection of 0.2 - 0.4 ng/well of recombinant Rat IL-6. For Neut: To yield one-half maximal inhibition [ND50] of the biological activity of Rat IL-6 (0.5 ng/ml), use at a concentration of 0.30 - 0.50 µg/ml. For WB: Use at a concentration of 0.1 - 0.2 µg/ml. In conjunction with compatible secondary reagents the detection limit for recombinant Rat IL-6 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR