Anti-CD43 Mouse Monoclonal Antibody [clone: MEM-59]

Supplier: GeneTex
GTX29088
89352-560EA 645.23 USD
89352-560
Anti-CD43 Mouse Monoclonal Antibody [clone: MEM-59]
Antibodies
Mouse monoclonal [MEM - 59] to CD43

FACS: Use at an assay dependant dilution. FuncS: Use at an assay dependant dilution. Activates T lymphocytes, this induces apoptosis in haemopoietic progenitor cells. IHC-P: Use at an assay dependant dilution. IP: Use at an assay dependant dilution. WB: Use at a dilution of 1/100. Use in non-reducing conditions. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD43
Clonality: Monoclonal
Clone: MEM-59
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR