Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-APOC2 Goat Polyclonal Antibody

Supplier: Genetex

GTX27618
89362-618EA 269.01 USD InStock
89362-618
Anti-APOC2 Goat Polyclonal Antibody
Antibodies
Goat Polyclonal antibody to Apolipoprotein C-II

Recommended Starting Dilutions:WB: Use at 1:5,000 - 1:10,000ELISA: Use at 1:5,000 - 1:10,000IHC: Use at 1:50 - 1:200IP: Use at 1:100

Type: Primary
Antigen: APOC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty