Anti-CAK1 Mouse Monoclonal Antibody [clone: MO-1.1]

Supplier: GeneTex
GTX23086
89351-856EA 425.89 USD
89351-856
Anti-CAK1 Mouse Monoclonal Antibody [clone: MO-1.1]
Antibodies
Mouse monoclonal [MO - 1.1] to CAK

Kinase assay: assy dependentFlow cytometry: assay dependent. Immunofluorescence: assay dependent. Immunoprecipitation (with protein A): use at a concentration of 2?g/mg protein lysate (native and denatured). Western blot: use at a concentration of 1 - 2?g/ml for 2 hours at RT. Immunohistochemistry (frozen sections): assay dependent. Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. By Western blot, this antibody detects a band of ~ 40 kDa, which corresponds to the predicted molecular weight of CAK.

Type: Primary
Antigen: CAK1
Clonality: Monoclonal
Clone: MO-1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR