Anti-BLIMP1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX89762
89367-010EA 607.87 USD
89367-010
Anti-BLIMP1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to PRDM1 / BLIMP1

Approx 100kDa band observed in Human Brain (cerebellum) lysates (calculated MW of 91.8kDa according to NP_001189.2 and of 76.8 according to NP_878911.1). Recommended concentration: 1-3µg/ml. This antibody is expected to recognize both reported isoforms (NP_001189.2 and NP_878911.1).

Type: Primary
Antigen: BLIMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR