Anti-PSCA Rabbit Polyclonal Antibody

GTX73982
89353-304EA 638.86 USD
89353-304
Anti-PSCA Rabbit Polyclonal Antibody
Polyclonal Antibody to PSCA ( Prostate Stem Cell Antigen )

This antibody may be diluted to a titer of 1:25-1:50 in an ABC method. However, depending on the staining conditions employed, we suggest that final dilution should be determined by the user. We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation conditions should be determined by the user. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining.

Type: Primary
Antigen: PSCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR