Anti-CP Goat Polyclonal Antibody

Supplier: GeneTex
GTX28749
89352-516EA 631.81 USD
89352-516
Anti-CP Goat Polyclonal Antibody
Antibodies
Goat polyclonal to Ceruloplasmin

Applications for immunohistochemistry have not been determined

Type: Primary
Antigen: CP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR