Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD79B Hamster Monoclonal Antibody

Supplier: Genetex

GTX75399
89365-794EA 246.6 USD InStock
89365-794
Anti-CD79B Hamster Monoclonal Antibody
Antibodies
Hamster monoclonal antibody [HM79-11] to CD79b

Type: Primary
Antigen: CD79B
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty