Anti-14-3-3 Mouse Monoclonal Antibody [clone: 3F7]

Supplier: GeneTex
GTX14110
89359-942EA 591.62 USD
89359-942
Anti-14-3-3 Mouse Monoclonal Antibody [clone: 3F7]
Antibodies
Members of the 14-3-3 family of proteins are highly conserved proteins, localized in neurons, and are axonally transported to the nerve terminals. They are also present, at lower levels, in various other eukaryotic tissues. 14-3-3 proteins appear to play important roles in a variety of signal transduction pathways, including those involved in cell cycle regulation and cell survival. Because 14-3-3 proteins bind to specific phosphoserine-containing sequences they are likely to have an important role in signaling pathways mediated by serine/threonine protein kinases. Evidence indicates 14-3-3 is required for Raf-1 kinase activity and phosphorylation amoung many other functions.

IP: Use at an assay dependent dilution. WB: Use at a concentration of 1 - 2 mg/ml. Predicted molecular weight: 30 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: 14-3-3
Clonality: Monoclonal
Clone: 3F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR