Anti-IgG Chicken Polyclonal Antibody

Supplier: GeneTex
GTX26863
89362-206EA 427.98 USD
89362-206
Anti-IgG Chicken Polyclonal Antibody
Antibodies
Biotin-conjugated Chicken anti-Human IgG polyclonal antibody

Recommended Starting Dilutions:1:20,000 - 1:400,000 for ELISA and Western blotting using enzyme-conjugated streptavidin 1:500 - 1:5,000 for enzyme immunohistochemistry on tissue sections 1:200 - 1:1,000 for Flow cytometry and ICC/IF

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR