Anti-IgG Chicken Polyclonal Antibody

Supplier: GeneTex
GTX77533
89353-768EA 212.31 USD
89353-768
Anti-IgG Chicken Polyclonal Antibody
Antibodies
HRP-conjugated Chicken anti-Human IgG (H+L) polyclonal antibody

Works in most applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR