Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Bovine RESPIRATORY SYNCYTIAL Virus Rabbit Polyclonal Antibody

Supplier: Genetex

GTX70321
89364-908EA 325.27 USD InStock
89364-908
Anti-Bovine RESPIRATORY SYNCYTIAL Virus Rabbit Polyclonal Antibody
Antibodies
Bovine Respiratory Syncytial Virus (BRSV) Fusion protein of the RB94 strain.

Type: Primary
Antigen: Bovine RESPIRATORY SYNCYTIAL Virus
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty