QMA 4.5CM PK100
Supplier: Whatman products (Cytiva)
Quartz Filters - Grade QM-A Circles, 45 mm 100/pk^ High-purity quartz microfiber filters are used for air sampling in acidic gases, stacks, flues and aerosols, particularly at high temperatures up to 500 Deg C and in PM-10 testing.